Dichloromethane ≥99.9% stabilized, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®

Warning

Synonyms: Methylene chloride

Stabilization: Stabilised with amylene 50-150 ppm

650463-20L-P2 650463-1L 650463-6X1L 650463-4L 650463-4X4L 650463-20L-N2
220084-760EA 316.92 USD
220084-760 220084-758 220084-763 220084-761 220084-762 220084-759
Dichloromethane ≥99.9% stabilized, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR