4-Amino-o-anisic acid ≥97%

Synonyms: 4-Amino-2-methoxybenzenecarboxylic acid

647624-25G 647624-5G
220084-917EA 61.38 USD
220084-917 220084-918
4-Amino-o-anisic acid ≥97%
4-Amino-o-anisic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00114479
CAS Number: 2486-80-8
EINECS: 219-632-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR