Molybdenum pentachloride, anhydrous

642452-2G 642452-10G
220084-139EA 137.08 USD
220084-139 220084-138
Molybdenum pentachloride, anhydrous
Molybdenum pentachloride
Formula: Cl₅Mo
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 194 °C
Density: 2.928 g/cm³ (20 °C)
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR