(3-Mercaptopropyl)triethoxysilane, Technical Grade

Synonyms: 3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane

63797-100ML-F 63797-25ML-F
220083-750EA 236.08 USD
220083-750 220083-751
(3-Mercaptopropyl)triethoxysilane, Technical Grade
(3-Mercaptopropyl)triethoxysilane
Formula: HSH₂CCH₂CH₂Si(OCH₂CH₃)₃
MW: 238.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00053924
CAS Number: 14814-09-6
EINECS: 238-883-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR