Manganese(II) carbonate hydrate

Synonyms: Manganous carbonate hydrate, Manganese carbonate hydrate

63539-250G 63539-1KG
220083-446EA 82.92 USD
220083-446 220083-445
Manganese(II) carbonate hydrate
Manganese(II) carbonate hydrate
Formula: CMnO₃·H₂O
MW: 132.96 g/mol
CAS Number: 34156-69-9
EINECS: 209-942-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR