1-Methylimidazolium hydrogen sulfate, Sigma-Aldrich®

Synonyms: 1-Methyl-1H-imidazole sulphate

59760-1KG-F 59760-100G-F
220081-350EA 1774.51 USD
220081-350 220081-349
1-Methylimidazolium hydrogen sulfate, Sigma-Aldrich®
1-Methylimidazolium hydrogen sulphate
Formula: C₄H₈N₂O₄S
MW: 180.18 g/mol
MDL Number: MFCD06798198
CAS Number: 681281-87-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR