3-Thiophenecarbonitrile

Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile

597449-25G 597449-5G
220081-562EA 156.15 USD
220081-562 220081-563
3-Thiophenecarbonitrile
3-Thiophenecarbonitrile
Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 82 °C (1013 hPa)
Density: 1.1956 g/cm³ (20 °C)
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR