Dimethyl itaconate

Synonyms: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

592498-100G 592498-25G
220081-126EA 453.38 USD
220081-126 220081-127
Dimethyl itaconate
Dimethyl itaconate
Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: 38 °C
Density: 1.1399 g/cm³ (14.7 °C)
MDL Number: MFCD00008446
CAS Number: 617-52-7
EINECS: 210-519-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR