Hafnium(IV) chloride

Synonyms: Hafnium tetrachloride

590592-5G 590592-25G
220081-033EA 323.08 USD
220081-033 220081-032
Hafnium(IV) chloride
Hafnium(IV) chloride
Formula: Cl₄Hf
MW: 320.3 g/mol
Melting Pt: 431.85 °C
MDL Number: MFCD00011034
CAS Number: 13499-05-3
EINECS: 236-826-5
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR