3-Phenyl-2-propyn-1-ol ≥96%

Synonyms: 3-Phenylprop-2-yn-1-ol

589306-5G 589306-25G
220078-193EA 42 USD
220078-193 220078-192
3-Phenyl-2-propyn-1-ol ≥96%
3-Phenyl-2-propyn-1-ol
Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00040914
CAS Number: 1504-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR