(4-Iodophenyl)acetonitrile ≥97%

Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile

587028-5G 587028-1G
220079-490EA 171.23 USD
220079-490 220079-489
(4-Iodophenyl)acetonitrile ≥97%
(4-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 56…57 °C
MDL Number: MFCD00060306
CAS Number: 51628-12-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR