Trifluoroacetic acid 0.1% (v/v) in acetonitrile for HPLC

Danger

574732-4X4L 574732-200L-LS 574732-20L 574732-18L 574732-4L
220080-168EA 4307.69 USD
220080-168 220080-165 220080-166 220080-164 220080-167
Trifluoroacetic acid 0.1% (v/v) in acetonitrile for HPLC
Trifluoroacetic acid
Formula: C₂HF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR