Indium(III) nitrate hydrate, Technical Grade

57135-U 57135-10G-F
220079-949EA 110.92 USD
220079-949 220079-948
Indium(III) nitrate hydrate, Technical Grade
Indium(III) nitrate hydrate
Formula: InN₃O₉·H₂O
MW: 318.85 g/mol
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR