(3-Chloropropyl)triethoxysilane

Synonyms: 3-Chloropropyltriethoxysilane, Triethoxy(3-chloropropyl)silane, 3-(Triethoxysilyl)propylchloride

569615-1L 569615-100ML
220079-771EA 93.46 USD
220079-771 220079-770
(3-Chloropropyl)triethoxysilane
(3-Chloropropyl)triethoxysilane
Formula: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
MW: 240.8 g/mol
MDL Number: MFCD00018985
CAS Number: 5089-70-3
EINECS: 225-805-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR