1-Bromo-3,5-dimethoxybenzene ≥97%

569313-25G 569313-5G
220079-673EA 104.89 USD
220079-673 220079-674
1-Bromo-3,5-dimethoxybenzene ≥97%
1-Bromo-3,5-dimethoxybenzene
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
MDL Number: MFCD06200685
CAS Number: 20469-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR