5-Iodoindole

Warning

563838-5G 563838-25G
220078-909EA 114.15 USD
220078-909 220078-908
5-Iodoindole
5-Iodoindole
Formula: C₈H₆IN
MW: 243.05 g/mol
MDL Number: MFCD00220065
CAS Number: 16066-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR