6-Aminobenzothiazole

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

562440-5G 562440-1G
220078-807EA 243.38 USD
220078-807 220078-806
6-Aminobenzothiazole
6-Aminobenzothiazole
Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD00015461
CAS Number: 533-30-2
EINECS: 208-559-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR