Nickel(II) bromide, anhydrous

Danger

Synonyms: Nickel dibromide

561142-10G 561142-2G
220078-696EA 446.15 USD
220078-696 220078-697
Nickel(II) bromide, anhydrous
Nickel(II) bromide
Formula: Br₂Ni
MW: 218.5 g/mol
Melting Pt: 963 °C
Density: 5.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 12,06585

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR