Zirconium(IV) tert-butanolate, Electronic Grade

Synonyms: Zirconium(IV) t-butoxide, Zirconium (IV) tert-butoxide, Zirconium (IV) tert-butylate

560030-5G 560030-25G
220078-526EA 403.08 USD
220078-526 220078-525
Zirconium(IV) tert-butanolate, Electronic Grade
Zirconium(IV) tert-butanolate
Formula: C₁₆H₃₆O₄Zr
MW: 383.68 g/mol
MDL Number: MFCD00075085
CAS Number: 2081-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR