3,5-Bis(trifluoromethyl)benzamide ≥97%

559997-1G 559997-5G
220078-522EA 30.8 USD
220078-522 220078-523
3,5-Bis(trifluoromethyl)benzamide ≥97%
3,5-Bis(trifluoromethyl)benzamide
Formula: C₉H₅F₆NO
MW: 257.14 g/mol
MDL Number: MFCD00015511
CAS Number: 22227-26-5
EINECS: 244-849-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR