5-Amino-2-bromopyridine

Warning

Synonyms: 6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine

552844-5G 552844-1G
220077-940EA 75.38 USD
220077-940 220077-939
5-Amino-2-bromopyridine
5-Amino-2-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD01646116
CAS Number: 13534-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR