8-Quinolinol hemisulfate salt

Warning

Synonyms: 8-Hydroxyquinoline hemisulphate, 8-Quinolinone hemisulphate, Oxine sulphate, Quinolin-8-ol hemisulphate, 8-Quinolinol sulphate, 8-Hydroxyquinolinium hemisulphate, Chinosol

55100-500G-F 55100-100G-F
220077-751EA 121.69 USD
220077-751 220077-750
8-Quinolinol hemisulfate salt
8-Quinolinol hemisulfate salt
Formula: C₁₈H₁₄N₂O₂·H₂SO₄
MW: 388.39 g/mol
Melting Pt: 177.5 °C
MDL Number: MFCD00013095
CAS Number: 134-31-6
EINECS: 205-137-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR