tert-Butyl acetoacetate ≥98%, Reagent Grade

Synonyms: tert-Butyl Acetoacetate

537594-500ML 537594-1L 537594-100ML 537594-4L
220076-417EA 200.31 USD
220076-417 220076-415 220076-414 220076-416
tert-Butyl acetoacetate ≥98%, Reagent Grade
tert-Butyl acetoacetate
Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: –38 °C
Density: 0.9756 g/cm³ (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00008811
CAS Number: 1694-31-1
EINECS: 216-904-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR