4-Nitro-o-toluonitrile ≥98%

Synonyms: 2-Methyl-4-nitrobenzonitrile, 4-Nitro-o-tolunitrile

537012-5G 537012-1G
220076-375EA 211.08 USD
220076-375 220076-374
4-Nitro-o-toluonitrile ≥98%
4-Nitro-o-toluonitrile
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03095378
CAS Number: 89001-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR