1-Butyl-3-methylimidazolium methyl sulfate, Sigma-Aldrich®

53177-100G-F 53177-1KG-F
220075-859EA 204 USD
220075-859 220075-860
1-Butyl-3-methylimidazolium methyl sulfate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium methyl sulphate
Formula: C₉H₁₈N₂O₄S
MW: 250.32 g/mol
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD03095437
CAS Number: 401788-98-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR