(2-Iodophenyl)acetic acid ≥97%

Warning

Synonyms: 2-(2-Iodophenyl)acetic acid

531472-25G 531472-5G
220075-305EA 115.23 USD
220075-305 220075-306
(2-Iodophenyl)acetic acid ≥97%
(2-Iodophenyl)acetic acid
Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
MDL Number: MFCD00046546
CAS Number: 18698-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR