Cetrimonium chloride

Danger

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

52366-50G 52366-10G
220074-949EA 513 USD
220074-949 220074-948
Cetrimonium chloride
Cetrimonium chloride
Formula: C₁₉H₄₂ClN
MW: 320 g/mol
Melting Pt: 232…233 °C
Density: 0.9 g/cm³ (20 °C)
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR