3-Hydroxyphenylboronic acid pinacol ester ≥97%

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

522562-1G 522562-5G
220074-510EA 59.12 USD
220074-510 220074-511
3-Hydroxyphenylboronic acid pinacol ester ≥97%
3-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR