1-Ethyl-3-methylimidazolium ethylsulfate, Sigma-Aldrich®

Synonyms: EMIM ESU

51682-1KG-F 51682-100G-F
220074-553EA 472 USD
220074-553 220074-552
1-Ethyl-3-methylimidazolium ethylsulfate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium ethylsulphate
Formula: C₈H₁₆N₂O₄S
MW: 236.29 g/mol
MDL Number: MFCD06798189
CAS Number: 342573-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR