1-Ethyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®

51059-1KG-F 51059-100G-F
220073-827EA 1230 USD
220073-827 220073-826
1-Ethyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium tetrachloroaluminate
Formula: C₆H₁₁AlCl₄N₂
MW: 279.96 g/mol
CAS Number: 80432-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR