4-Ethylphenylboronic acid

Warning

Synonyms: (4-Ethylphenyl)boronic acid, (4-Ethylphenyl)metaboric acid, 4-Ethylbenzeneboronic acid, Dihydroxy(4-ethylphenyl)borane

499536-5G 499536-25G
220073-527EA 20.83 USD
220073-527 220073-526
4-Ethylphenylboronic acid
4-Ethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00859377
CAS Number: 63139-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR