Sodium-L(+)-glutamate monohydrate

49621-1KG 49621-250G
220073-257EA 96.17 USD
220073-257 220073-258
Sodium-L(+)-glutamate monohydrate
Sodium-L(+)-glutamate monohydrate
Formula: C₅H₈NNaO₄·H₂O
Melting Pt: 225 - 240 °C
Flash Pt: ≥ 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR