2-Chloro-5-methylpyridine

Warning

Synonyms: 2-Chloro-5-picoline

495328-25ML 495328-5ML
220073-206EA 168 USD
220073-206 220073-207
2-Chloro-5-methylpyridine
2-Chloro-5-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 97 °C (30 mmHg)
Density: 1.169 g/cm³ (20 °C)
MDL Number: MFCD00792460
CAS Number: 18368-64-4
EINECS: 418-050-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR