Cobalt (III) acetylacetonate

Synonyms: Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)

494534-25G 494534-5G
220072-327EA 269.23 USD
220072-327 220072-328
Cobalt (III) acetylacetonate
Cobalt (III) acetylacetonate
Formula: C₁₅H₂₁CoO₆
MW: 356.26 g/mol
MDL Number: MFCD00013488
CAS Number: 21679-46-9
EINECS: 244-527-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR