Titanium(II) oxide, deposition grade

Synonyms: Titanium oxide, Titanium monoxide

481041-50G 481041-10G
220071-890EA 223.08 USD
220071-890 220071-889
Titanium(II) oxide, deposition grade
Titanium(II) oxide
Formula: OTi
MW: 63.88 g/mol
Boiling Pt: 3660 °C (1013 hPa)
Melting Pt: 1750 °C
Density: 4.88 g/cm³ (20 °C)
MDL Number: MFCD00036279
CAS Number: 12137-20-1
EINECS: 235-236-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR