2,6-Dichlorophenyl isothiocyanate ≥97%

Danger

Synonyms: 1,3-Dichloro-2-isothiocyanatobenzene

475890-5G 475890-1G
220071-400EA 97.35 USD
220071-400 220071-399
2,6-Dichlorophenyl isothiocyanate ≥97%
2,6-Dichlorophenyl isothiocyanate
Formula: C₇H₃Cl₂NS
MW: 204.08 g/mol
Boiling Pt: 100 °C (0.3 -rr)
Melting Pt: 44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041056
CAS Number: 6590-95-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR