1-Hexanol, anhydrous ≥99%

Warning

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

471402-2L 471402-1L 471402-100ML
220070-020EA 93.23 USD
220070-020 220070-019 220070-018
1-Hexanol, anhydrous ≥99%
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.8322 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
Merck Index: 13,04715
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR