Yttrium(III) chloride hexahydrate

Warning

464317-25G 464317-100G
220069-727EA 100.46 USD
220069-727 220069-726
Yttrium(III) chloride hexahydrate
Yttrium(III) chloride hexahydrate
Formula: YCl₃·6H₂O
MW: 303.36 g/mol
Boiling Pt: 1507 °C (1013 hPa)
Melting Pt: 153 °C
Density: 2.18 g/cm³ (20 °C)
MDL Number: MFCD00149941
CAS Number: 10025-94-2
EINECS: 233-801-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR