2,4-Dimethylphenylhydrazinium chloride ≥97%

Synonyms: 1-(2,4-Dimethylphenyl)hydrazine hydrochloride, 2,4-Dimethylphenylhydrazine hydrochloride

462888-5G 462888-25G
220069-447EA 109.85 USD
220069-447 220069-446
2,4-Dimethylphenylhydrazinium chloride ≥97%
2,4-Dimethylphenylhydrazinium chloride
Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
MDL Number: MFCD00013381
CAS Number: 60480-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR