1-Pentyl-4-ethynylbenzene

Synonyms: 4-N-Pentylphenylacetylene, 4-n-Pentylphenylacetylene, (4-Pentylphenyl)acetylene, 1-Ethynyl-4-pentylbenzene, (4-Pentylphenyl)ethyne

457477-1G 457477-10G
220069-327EA 42.86 USD
220069-327 220069-326
1-Pentyl-4-ethynylbenzene
1-Pentyl-4-ethynylbenzene
Formula: C₁₃H₁₆
MW: 172.27 g/mol
MDL Number: MFCD00173884
CAS Number: 79887-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR