Lithium iodide hydrate ≥99.999%

452327-25G 452327-5G
220067-202EA 358.46 USD
220067-202 220067-203
Lithium iodide hydrate ≥99.999%
Lithium iodide hydrate
Formula: ILi·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149769
CAS Number: 17023-25-5
EINECS: 233-822-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR