Titanium(II) chloride, anhydrous ≥99.98% (trace metals basis), powder

Danger

Synonyms: Titanium(II) chloride, Dichlorotitanium

451738-1G 451738-5G
220068-686EA 403.08 USD
220068-686 220068-687
Titanium(II) chloride, anhydrous ≥99.98% (trace metals basis), powder
Titanium(II) chloride
Formula: TiCl₂
MW: 118.77 g/mol
Density: 5.38 g/cm³ (20 °C)
MDL Number: MFCD00054968
CAS Number: 10049-06-6
EINECS: 233-165-4
UN: 3200
ADR: 4.2,I
Merck Index: 12,08355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR