Scandium(III) chloride, anhydrous

451266-5G 451266-1G
220067-682EA 1800 USD
220067-682 220067-681
Scandium(III) chloride, anhydrous
Scandium(III) chloride
Formula: Cl₃Sc
MW: 151.31 g/mol
Melting Pt: 940 °C
Density: 1.67 g/cm³ (940 °C)
MDL Number: MFCD00011221
CAS Number: 10361-84-9
EINECS: 233-799-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR