Chromium(III) chloride, anhydrous

Warning

Synonyms: Chromium trichloride

450790-25G 450790-5G
220068-572EA 681.54 USD
220068-572 220068-573
Chromium(III) chloride, anhydrous
Chromium(III) chloride
Formula: Cl₃Cr
MW: 158.35 g/mol
Melting Pt: 1152 °C
Density: 2.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 12,02278

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR