Strontium ≥99.99%

441899-5G 441899-25G
220065-982EA 1769.23 USD
220065-982 220067-774
Strontium ≥99.99%
Strontium
Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1151 °C (1013 hPa)
Melting Pt: 570…660 °C
Density: 2.38 g/cm³ (770 °C)
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3,II
Merck Index: 12,08994

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR