(3-Chloropropyl)trimethoxysilane

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

440183-100ML 440183-1L
220067-572EA 80.68 USD
220067-572 220067-573
(3-Chloropropyl)trimethoxysilane
(3-Chloropropyl)trimethoxysilane
Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MW: 198.72 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.082 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000997
CAS Number: 2530-87-2
EINECS: 219-787-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR