β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt

43420-1G 43420-100MG 43420-5G
220067-060EA 276.92 USD
220067-060 220067-059 220067-061
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR