5-Bromo-2-furaldehyde

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

433985-1G 433985-10G
220067-033EA 153.69 USD
220067-033 220067-032
5-Bromo-2-furaldehyde
5-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 201…203 °C (1013 hPa)
Melting Pt: 74…76 °C
MDL Number: MFCD00159501
CAS Number: 1899-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR