3-(Trifluoromethyl)phenylboronic acid

Synonyms: (3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane

432032-5G 432032-1G
220066-918EA 112.15 USD
220066-918 220066-917
3-(Trifluoromethyl)phenylboronic acid
3-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
MDL Number: MFCD00151854
CAS Number: 1423-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR