Cesium bromide, anhydrous ≥99.999%

Warning

429392-1G 429392-10G
220066-667EA 66.46 USD
220066-667 220066-666
Cesium bromide, anhydrous ≥99.999%
Cesium bromide
Formula: BrCs
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 636 °C
Density: 4.46 g/cm³ (20 °C)
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 13,02019

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR